Microtubes, Prepared
Microtubes, prepared, haematology potassium EDTA, with screw cap closure, 1,3 ml, 100/case
Artikelnr. SARS41.1395.100
Marketsource
Anbieter: SARSTEDT
Springe zu:
Product Details & Documents
PP, translucent with PP screw cap.
- Type of preparation: Trisodium citrate solution
- With paper label
- Graduated
- Recommended centrifugation: 1800×g
With screw cap or push-on cap.
Spezifikationen
- Verpackt:100/case
- Kapazität:1,3 ml
- Bezeichnung:Microtubes, prepared, haematology potassium EDTA, with screw cap closure
- Seq:7