Connectors, PP
Connector, straight, female luer lock/tubing nipple, 6,4 mm
Artikelnr. INFIPP-LFS64
Anbieter: PIEPER FILTER
Springe zu:
Product Details & Documents
Connectors, made of PP, natural.
- Good chemical resistance
- Good temperature resistance
Spezifikationen
- Schlauch Innen-Ø:6,4 mm
- Bezeichnung:Connector, straight, female luer lock/tubing nipple
- Seq:12