Stoppers, for test tubes
Blau, 11 - 12 mm
Artikelnr. RATI351490B
Anbieter: RATIOLAB
Springe zu:
Product Details & Documents
PE.
- Very tight seal due to two sealing rings
- Available in natural and four more colors
Spezifikationen
- Ø unten:11 - 12 mm
- Farbe:Blau
- Seq:02