AuftragsverfolgungKontakt
Drum stands, FB
Drum stands, FB
Drum stands, FB 3, fire-galvanised, capacity 3×60 l drum, 1230×650×415 mm
  LACOP21-1703-A
Drum stands, FB
  LACOP21-1703-A
Drum stands, FB
Drum stands, FB 3, fire-galvanised, capacity 3×60 l drum, 1230×650×415 mm
1 * 1 ST
Drum stands with fixed drum supports for 60 to 200 L drums, completely galvanised. Tilted support onwards for easy drum emptying.
These drum stands can be combined with compact sump pallets, water protection pallets, hazardous substance stations, floor safety elements and drum carts.
  • Bezeichnung:
    Drum stands, FB 3, fire-galvanised, capacity 3×60 l drum
  • Seq:
    05
  • B×T×H:
    1230×650×415 mm