Express Eingabe
Austria
AuftragsverfolgungKontakt
Polystyrene, analytical standards for GPC | Avantor
Polystyrene, analytical standards for GPC
Mw 900000 (typical), 1 g
  327808-1G
 :   Merck
Polystyrene, analytical standards for GPC
  327808-1G
 :  Merck
 :  
Polystyrene, analytical standards for GPC
Polystyrene, analytical standards for GPC
Mw 900000 (typical), 1 g
 :  
115,00 €
1 * 1 g
Zertifizierung: Exact values can be taken from the accompanying certificate of analysis
Achtung! Bitte beachten: Storage temperature: 2...8 °C. Limited shelf life, expiry date on the label.
  • VE:
    1 g
  • Bezeichnung:
    Mw 900000 (typical)
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.