Polystyrene, analytical standards for GPC
Mw 900000 (typical), 1 g
327808-1G
: Merck
Zertifizierung: Exact values can be taken from the accompanying certificate of analysis
Achtung! Bitte beachten: Storage temperature: 2...8 °C. Limited shelf life, expiry date on the label.
- VE:1 g
- Bezeichnung:Mw 900000 (typical)