Express Eingabe
Austria
AuftragsverfolgungKontakt
Polystyrene, analytical standards for GPC

Polystyrene, analytical standards for GPC

Mw 200000 (typical), 1 g
Artikelnr. 327786-1G
Anbieter:   Merck
Polystyrene, analytical standards for GPC
Artikelnr. 327786-1G
Anbieter:  Merck
CAS Nummer:  
Polystyrene, analytical standards for GPC
Polystyrene, analytical standards for GPC

Mw 200000 (typical), 1 g

CAS Nummer:  
Preis
119,00 €
Einloggen für Verfügbarkeit
1 * 1 g
Product Details & Documents

Certifications: Exact values can be taken from the accompanying certificate of analysis

Caution: Storage temperature: 2...8 °C. Limited shelf life, expiry date on the label.

Spezifikationen

Spezifikationen

  • VE:
    1 g
  • Bezeichnung:
    Mw 200000 (typical)

Spezifikationen

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.