Filling cannulae
Filling cannulae, G 20, 50 mm
Catalog # MURRM720/03
Supplier: MURRAY S
Jump to:
Product Details & Documents
Filling cannulae with blunt end and Luer lock fitting.
: Different sizes and gauges are available.
Specifications
- Gauge:G 20
- Bezeichnung:Filling cannulae
- Länge:50 mm
- Seq:005