Filling cannulae
Filling cannulae, G 18, 150 mm
Artikelnr. MURRM718/08
Anbieter: MURRAY S
Springe zu:
Product Details & Documents
Filling cannulae with blunt end and Luer lock fitting.
Ordering information: Different sizes and gauges are available.
Documents
MURRM714^06_TDS_EN
Datasheet
Spezifikationen
- Gauge:G 18
- Bezeichnung:Filling cannulae
- Länge:150 mm
- Seq:019