Filling cannulae
Filling cannulae, G 14, 76 mm
Artikelnr. MURRM714/05
Anbieter: MURRAY S
Springe zu:
Product Details & Documents
Filling cannulae with blunt end and Luer lock fitting.
Ordering information: Different sizes and gauges are available.
Documents
MURRM714^06_TDS_EN
Datasheet
Spezifikationen
- Gauge:G 14
- Bezeichnung:Filling cannulae
- Länge:76 mm
- Seq:007