Polystyrene standards, GPC/SEC columns calibration
Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,11, quantities 01 g, 1
Artikelnr. VARIPL2012-0001
Anbieter: VARIAN
Polystyrene standards, GPC/SEC columns calibration
Artikelnr. VARIPL2012-0001
Anbieter: VARIAN
CAS Nummer:
Springe zu:
Product Details & Documents
Polystyrene standards for many organic solvents either for conventional GPC column calibration or for calibrating light scattering and viscosity detectors that provide equidistant calibration points for greater accuracy.
- Compatible with most organic solvents
- Certificate of analysis meets international protocols
- Calibration capability for virtually all applications
Zertifikate
Suche nach ChargennummerSpezifikationen
Spezifikationen
- VE:1
- Bezeichnung:Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,11, quantities 01 g