AuftragsverfolgungKontakt
Polystyrene standards, GPC/SEC columns calibration
Polystyrene calibration kit, S-H2-10, 10×0,5 g, 1
Catalog # VARIPL2010-0104
Supplier:   VARIAN
Polystyrene standards, GPC/SEC columns calibration
Catalog # VARIPL2010-0104
Supplier:  VARIAN
CAS Number:  
Polystyrene standards, GPC/SEC columns calibration
Polystyrene calibration kit, S-H2-10, 10×0,5 g, 1
CAS Number:  
Price
1 237,83 €
Log in for availability
1 * 1 ST
Product Details & Documents
Polystyrene standards for many organic solvents either for conventional GPC column calibration or for calibrating light scattering and viscosity detectors that provide equidistant calibration points for greater accuracy.
  • Compatible with most organic solvents
  • Certificate of analysis meets international protocols
  • Calibration capability for virtually all applications
Specifications
Specifications
  • VE:
    1
  • Bezeichnung:
    Polystyrene calibration kit, S-H2-10, 10×0,5 g
Specifications