AuftragsverfolgungKontakt
Polystyrene standards, GPC/SEC columns calibration
Polystyrene calibration kit, S-M2-10, 10×0,5 g, 1
Artikelnr. VARIPL2010-0102
Anbieter:   VARIAN
Polystyrene standards, GPC/SEC columns calibration
Artikelnr. VARIPL2010-0102
Anbieter:  VARIAN
CAS Nummer:  
Polystyrene standards, GPC/SEC columns calibration
Polystyrene calibration kit, S-M2-10, 10×0,5 g, 1
CAS Nummer:  
Preis
1 104,84 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Polystyrene standards for many organic solvents either for conventional GPC column calibration or for calibrating light scattering and viscosity detectors that provide equidistant calibration points for greater accuracy.
  • Compatible with most organic solvents
  • Certificate of analysis meets international protocols
  • Calibration capability for virtually all applications
Spezifikationen
Spezifikationen
  • VE:
    1
  • Bezeichnung:
    Polystyrene calibration kit, S-M2-10, 10×0,5 g