Calibration standards for size exclusion chromatography
Standard SM-105, 0,5 g × 10 standards, with MW range 1200 - 3800000, PS, 1
Artikelnr. SHOWF8602105
Anbieter: Resonac
Calibration standards for size exclusion chromatography
Artikelnr. SHOWF8602105
Anbieter: Resonac
CAS Nummer:
Springe zu:
Product Details & Documents
Dokumente
Spezifikationen
Spezifikationen
- VE:1
- Bezeichnung:Standard SM-105, 0,5 g × 10 standards, with MW range 1200 - 3800000, PS
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.