Calibration standards for size exclusion chromatography
Standard P-82, 0,2 g × 8 standards, with MW range 5000 - 800000, pullulan, 1
Anbieter: Resonac
Calibration standards for size exclusion chromatography
Artikelnr. SHOWF8400000
Anbieter: Resonac
CAS Nummer:
Springe zu:
Product Details & Documents
Dokumente
Spezifikationen
Spezifikationen
- VE:1
- Bezeichnung:Standard P-82, 0,2 g × 8 standards, with MW range 5000 - 800000, pullulan
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.