AuftragsverfolgungKontakt
OVS sampling tubes, ORBO™
ORBO™ 65P (OVS) Amberlite® XAD®-4 specially cleaned, 160/80 mg RC,GFF,F,F (specially cleaned) separators, OSHA Versatile Sampler (OVS), O.D.×L: 13-8×75 mm, package of 10 ea
Artikelnr. 20029-U
Anbieter:   Merck
OVS sampling tubes, ORBO™
Artikelnr. 20029-U
Anbieter:  Merck
OVS sampling tubes, ORBO™
ORBO™ 65P (OVS) Amberlite® XAD®-4 specially cleaned, 160/80 mg RC,GFF,F,F (specially cleaned) separators, OSHA Versatile Sampler (OVS), O.D.×L: 13-8×75 mm, package of 10 ea
Preis
342,00 €
Einloggen für Verfügbarkeit
1 * 1 Pack.
Product Details & Documents
The OSHA Versatile Sampler (OVS) is a unique tube design with the adsorbent and filter combined in one tube for collection of both vapors and aerosols.
  • Packed with specially cleaned low background porous polymers
  • Most common sampling rate is 1 L/min to obtain a sample volume from 60 to 480 L
It′s most common use is for sampling pesticides but can also be used for other applications.
Spezifikationen
  • Bezeichnung:
    ORBO™ 65P (OVS) Amberlite® XAD®-4 specially cleaned, 160/80 mg RC,GFF,F,F (specially cleaned) separators, OSHA Versatile Sampler (OVS), O.D.×L: 13-8×75 mm, package of 10 ea
  • Seq:
    01