Solid phase extraction cartridges, HyperSep™ SAX
Kartusche, SAX, 40 - 60 µm, 25 ml, 5000 mg
HYPE60108-714
Strong anion exchange sorbent for extraction of weak acids.
- Extracts negatively charged compounds from both aqueous and non-aqueous solutions
- Ideally suited for the extraction of weak acids, e.g. carboxylic acids
- Amine group masks the effect of the carbon chain of the functional group
- Selectivity can be tuned by selection of buffer
- Säulenform:Kartusche
- Partikelgröße:40 - 60 µm
- Kapazität:25 ml
- Seq:08
- Bettgewicht:5000 mg
- Phase:SAX



