Solid phase extraction cartridges, HyperSep™ SAX
Kartusche, SAX, 40 - 60 µm, 1 ml, 50 mg
HYPE60108-417
Strong anion exchange sorbent for extraction of weak acids.
- Extracts negatively charged compounds from both aqueous and non-aqueous solutions
- Ideally suited for the extraction of weak acids, e.g. carboxylic acids
- Amine group masks the effect of the carbon chain of the functional group
- Selectivity can be tuned by selection of buffer
- Säulenform:Kartusche
- Partikelgröße:40 - 60 µm
- Kapazität:1 ml
- Seq:01
- Bettgewicht:50 mg
- Phase:SAX
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



