Washers, BOLA
M 6 thread, 12 mm
Catalog # BOHLH1126-22
Supplier: Bohlender
Jump to:
Product Details & Documents
PTFE, white.
- Temperature resistance: from −200 to +250 °C
- Universal chemical resistance
- FDA conform
- Similar to DIN 125-1
Specifications
- Ø:12 mm
- Bezeichnung:M 6 thread