AuftragsverfolgungKontakt
Control valves, BOLA
Control valves, BOLA
Control valve with thread M 18×2, for tubing outer Ø 10 mm, PTFE
Artikelnr. BOHLE672-56
Anbieter:   Bohlender
Control valves, BOLA
Artikelnr. BOHLE672-56
Anbieter:  Bohlender
Control valves, BOLA
Control valves, BOLA
Control valve with thread M 18×2, for tubing outer Ø 10 mm, PTFE
Preis
395,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Body made of PTFE with 2 connections and nuts made of glass-fibre reinforced PTFE for connecting tubing or tube.
  • Volume flow can be regulated manually
  • Suitable for panel mounting
  • Easy disassembly for cleaning
  • Universal chemical resistance, the flowing product is only exposed to PTFE
Spezifikationen
  • Bezeichnung:
    Control valve with thread M 18×2, for tubing outer Ø 10 mm, PTFE