Operating scissors
Straight, sharp/blunt, 115 mm
Artikelnr. HAMML-HSB084-11
Anbieter: HAMMACHER KARL
Springe zu:
Product Details & Documents
These operating scissors made from stainless steel are suitable for medical and clinical applications.
Spezifikationen
- Typ:Straight, sharp/blunt
- Länge:115 mm
- Seq:03