AuftragsverfolgungKontakt
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Carbon-filtered pressure relief vent kit, incl. activated carbon filter, for ducting to outside or room; may be used with 5238500 HEPA-filtered pressure relief vent kit
Artikelnr. LBCP5241200
Anbieter:   LABCONCO
Accessories for glove boxes, Precise™
Artikelnr. LBCP5241200
Anbieter:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Carbon-filtered pressure relief vent kit, incl. activated carbon filter, for ducting to outside or room; may be used with 5238500 HEPA-filtered pressure relief vent kit
Preis
2 730,00 €
Einloggen für Verfügbarkeit
1 * 1 KIT
Product Details & Documents
Spezifikationen
  • Bezeichnung:
    Carbon-filtered pressure relief vent kit, incl. activated carbon filter, for ducting to outside or room; may be used with 5238500 HEPA-filtered pressure relief vent kit
  • Seq:
    12