Accessories for glove boxes, Precise™
HEPA-filtered pressure relief vent kit, with 99,99% efficient HEPA filter; for ducting to the outside or room; may be used with 5241200 carbon-filtered vent kit
Artikelnr. LBCP5238500
Anbieter: LABCONCO
Springe zu:
Product Details & Documents
Spezifikationen
- Bezeichnung:HEPA-filtered pressure relief vent kit, with 99,99% efficient HEPA filter; for ducting to the outside or room; may be used with 5241200 carbon-filtered vent kit
- Seq:11