AuftragsverfolgungKontakt
Accessories for glove boxes, Protector® | Avantor
Accessories for glove boxes, Protector®
Universal exterior glove port covers, molded neoprene, incl. 2 clamps, may be placed over glove ports to facilitate and expedite the evacuation/fill operation and preserve inert atmosphere
Artikelnr. LBCP5242200
Anbieter:   LABCONCO
Accessories for glove boxes, Protector®
Artikelnr. LBCP5242200
Anbieter:  LABCONCO
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Universal exterior glove port covers, molded neoprene, incl. 2 clamps, may be placed over glove ports to facilitate and expedite the evacuation/fill operation and preserve inert atmosphere
Preis
261,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Dokumente
Spezifikationen
  • Bezeichnung:
    Universal exterior glove port covers, molded neoprene, incl. 2 clamps, may be placed over glove ports to facilitate and expedite the evacuation/fill operation and preserve inert atmosphere
  • Seq:
    13