Accessories for glove boxes, Protector®
Universal exterior glove port covers, molded neoprene, incl. 2 clamps, may be placed over glove ports to facilitate and expedite the evacuation/fill operation and preserve inert atmosphere
Artikelnr. LBCP5242200
Anbieter: LABCONCO
Springe zu:
Product Details & Documents
Dokumente
LBCP5242200_TDS_EN
Technisches Datenblatt
Spezifikationen
- Bezeichnung:Universal exterior glove port covers, molded neoprene, incl. 2 clamps, may be placed over glove ports to facilitate and expedite the evacuation/fill operation and preserve inert atmosphere
- Seq:13
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.