Accessories for glove boxes, Precise™
Viewing window, acrylic (749×396×12,7 mm) with dry powder epoxy-coated steel frame, replaces standard laminated glass window
Artikelnr. LBCP5236100
Anbieter: LABCONCO
Springe zu:
Product Details & Documents
Spezifikationen
- Bezeichnung:Viewing window, acrylic (749×396×12,7 mm) with dry powder epoxy-coated steel frame, replaces standard laminated glass window
- Seq:36