AuftragsverfolgungKontakt
Accessories for glove boxes, Precise™
Moisture monitor/hygrometer for placement inside the main chamber to continuously monitor moisture levels and absolute moisture content in gas (0 - 10000 ppm); consists of chemically inert ceramic technology moisture sensor, sampling block, digital display monitor, sampling pump, switching power supply and on/off switch, for 230 V units
Catalog # LBCP5244901
Supplier:   LABCONCO
Accessories for glove boxes, Precise™
Catalog # LBCP5244901
Supplier:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Moisture monitor/hygrometer for placement inside the main chamber to continuously monitor moisture levels and absolute moisture content in gas (0 - 10000 ppm); consists of chemically inert ceramic technology moisture sensor, sampling block, digital display monitor, sampling pump, switching power supply and on/off switch, for 230 V units
Price
14 270,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Bezeichnung:
    Moisture monitor/hygrometer for placement inside the main chamber to continuously monitor moisture levels and absolute moisture content in gas (0 - 10000 ppm); consists of chemically inert ceramic technology moisture sensor, sampling block, digital display monitor, sampling pump, switching power supply and on/off switch, for 230 V units
  • Seq:
    14