Accessories for glove boxes, Precise™
Pressure relief valve vent kit for exhaustig the pressure relief valve during overpressurization, incl. 3,9" OD × 3,8" ID thimble connection, gasket, two elbows, four clamps and 13" long tubing (Ductwork for outside venting and 5230600 Pressure Relief Valve are required but not included)
Artikelnr. LBCP5242700
Anbieter: LABCONCO
Springe zu:
Product Details & Documents
Spezifikationen
- Bezeichnung:Pressure relief valve vent kit for exhaustig the pressure relief valve during overpressurization, incl. 3,9" OD × 3,8" ID thimble connection, gasket, two elbows, four clamps and 13" long tubing (Ductwork for outside venting and 5230600 Pressure Relief Valve are required but not included)
- Seq:13