AuftragsverfolgungKontakt
Accessories for glove boxes, Precise™
Tissue holder, epoxy-coated steel, accommodates a box of tissues in the upside down or upright position; opening in the holder permits wipes to be dispensed from the bottom
Artikelnr. LBCP3927800
Anbieter:   LABCONCO
Accessories for glove boxes, Precise™
Artikelnr. LBCP3927800
Anbieter:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Tissue holder, epoxy-coated steel, accommodates a box of tissues in the upside down or upright position; opening in the holder permits wipes to be dispensed from the bottom
Preis
160,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Spezifikationen
  • Bezeichnung:
    Tissue holder, epoxy-coated steel, accommodates a box of tissues in the upside down or upright position; opening in the holder permits wipes to be dispensed from the bottom
  • Seq:
    42