AuftragsverfolgungKontakt
Accessories for glove boxes, Precise™
Electrical pass-through kit, provides a leak-tight connection for data transfer to a printer or computer; incl. sealed 9 pin connector (D-subminiature with male pins on one side and female pins on the other) and 10' cord; requires installation and drill hole; Installation Kit 5241100 is recommended
Artikelnr. LBCP5075619
Anbieter:   LABCONCO
Accessories for glove boxes, Precise™
Artikelnr. LBCP5075619
Anbieter:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Electrical pass-through kit, provides a leak-tight connection for data transfer to a printer or computer; incl. sealed 9 pin connector (D-subminiature with male pins on one side and female pins on the other) and 10' cord; requires installation and drill hole; Installation Kit 5241100 is recommended
Preis
292,00 €
Einloggen für Verfügbarkeit
1 * 1 KIT
Product Details & Documents
Spezifikationen
  • Bezeichnung:
    Electrical pass-through kit, provides a leak-tight connection for data transfer to a printer or computer; incl. sealed 9 pin connector (D-subminiature with male pins on one side and female pins on the other) and 10' cord; requires installation and drill hole; Installation Kit 5241100 is recommended
  • Seq:
    32