Pulvertrichter
160 mm, 31 mm, 25 mm
Artikelnr. SEMA0213
Anbieter: SEMADENI
Springe zu:
Product Details & Documents
Funnels, made of PP, translucent. Used for transferring powders. Also suitable for liquids.
- Parallel stem minimises powder bridging
- Very good chemical and temperature resistance
- Autoclavable
Spezifikationen
- Trichter A-Ø:160 mm
- Stiel A-Ø:31 mm
- Stiel Länge:25 mm



