Schwefel ≥99%, Blume
Glass bottle for solids, 500 g
Anbieter: Thermo Fisher Scientific
Springe zu:
Product Details & Documents
- CAS Nummer:7704-34-9
- Formel:S
- MDL-Nummer:MFCD00085316
- Molekulargewicht:32.07 g/mol
- Merck-Index:13,09059
- Siedepunkt:445 °C (1013 hPa)
- Schmelzpunkt:113…119 °C
- Dichte:2,36 g/cm³ (20 °C)
- Blitzpunkt:168 °C
- Lagertemperatur:Raumtemperatur
- EINECS:231-722-6
- UN:1350
- ADR:4.1,III
Dokumente
Spezifikationen
Spezifikationen
- Packung:Glass bottle for solids
- VE:500 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



