Forceps
Scharf, gekrümmt, 115 mm
Catalog # 232-0200
Supplier: MURRAY S
Jump to:
Product Details & Documents
Stainless steel. Dissecting forceps with guide pin, which helps to keep the fine tips aligned.
- Sharp, narrow flutes
- Curved
Specifications
- Typ:Scharf, gekrümmt
- Länge:115 mm
- Cat. No.:232-0200