Express Eingabe
Austria
AuftragsverfolgungKontakt
Solid phase extraction cartridges, HyperSep™ SCX

Solid phase extraction cartridges, HyperSep™ SCX

Kartusche, SCX, 40 - 60 µm, 6 ml, 500 mg
Catalog # HYPE60108-520
Solid phase extraction cartridges, HyperSep™ SCX
Catalog # HYPE60108-520
Solid phase extraction cartridges, HyperSep™ SCX
Solid phase extraction cartridges, HyperSep™ SCX

Kartusche, SCX, 40 - 60 µm, 6 ml, 500 mg

Price
297,00 €
Log in for availability
1 * 30 ST
Product Details & Documents

A strong cation exchange sorbent for extraction of charged basic compounds.

  • Benzene ring provides potential for nonpolar interactions
  • Low pKa value sorbent
Specifications
  • Säulenform:
    Kartusche
  • Partikelgröße:
    40 - 60 µm
  • Kapazität:
    6 ml
  • Seq:
    05
  • Bettgewicht:
    500 mg
  • Phase:
    SCX
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.