Solid phase extraction cartridges, HyperSep™ SAX
Kartusche, SAX, 40 - 60 µm, 3 ml, 500 mg
Artikelnr. HYPE60108-521
Anbieter: Thermo Fisher Scientific
Solid phase extraction cartridges, HyperSep™ SAX
Artikelnr. HYPE60108-521
Anbieter: Thermo Fisher Scientific
Springe zu:
Product Details & Documents
Strong anion exchange sorbent for extraction of weak acids.
- Extracts negatively charged compounds from both aqueous and non-aqueous solutions
- Ideally suited for the extraction of weak acids, e.g. carboxylic acids
- Amine group masks the effect of the carbon chain of the functional group
- Selectivity can be tuned by selection of buffer
Spezifikationen
- Säulenform:Kartusche
- Partikelgröße:40 - 60 µm
- Kapazität:3 ml
- Seq:04
- Bettgewicht:500 mg
- Phase:SAX



