AuftragsverfolgungKontakt
LC-MS nitrogen generators | Avantor
LC-MS nitrogen generators
N2-35 Mid Flow, without air compressor, 20 l/min at 100 psi; 25 l/min at 125 psi; 30 l/min at 100 psi; 37 l/min at 125 psi; 46 l/min at 100 psi; 57 l/min at 125 psi, 450×410×1300 mm
Artikelnr. PERCN9306211
Anbieter:   PerkinElmer
LC-MS nitrogen generators
Artikelnr. PERCN9306211
Anbieter:  PerkinElmer
LC-MS nitrogen generators
N2-35 Mid Flow, without air compressor, 20 l/min at 100 psi; 25 l/min at 125 psi; 30 l/min at 100 psi; 37 l/min at 125 psi; 46 l/min at 100 psi; 57 l/min at 125 psi, 450×410×1300 mm
Preis
40 250,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Nitrogen generators are designed to supply single or multiple LC-MS instruments with dry nitrogen at purities of 98 – 99,5%. The generator can also be used for solvent evaporation as well as supplying dry nitrogen to analytical instruments.
Spezifikationen
  • Förderleistung:
    20 l/min at 100 psi; 25 l/min at 125 psi; 30 l/min at 100 psi; 37 l/min at 125 psi; 46 l/min at 100 psi; 57 l/min at 125 psi
  • B×T×H:
    450×410×1300 mm
  • Typ:
    N2-35 Mid Flow, without air compressor