Rectangular spacers for SpectrumGX
Spacers, lead, rectangular, 0,025 mm thickness for semi-demountable cell, SpectrumGX
Artikelnr. PERCL1270301
Anbieter: PerkinElmer
Springe zu:
Product Details & Documents
PTFE spacers can be used for the analysis of less viscous samples.
Spezifikationen
- Für:SpectrumGX
- Bezeichnung:Spacers, lead, rectangular, 0,025 mm thickness for semi-demountable cell
- Seq:01