Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Trypanblau

10 g
Catalog # ROTHCN76.1
Marketsource
Supplier:   Roth Carl
Trypanblau
Catalog # ROTHCN76.1
Supplier:  Roth Carl
Marketsource
CAS Number:  72-57-1
Trypanblau

10 g

CAS Number:  72-57-1
Price
33,20 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, AzidinBlue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)
  • CAS Number:72-57-1
  • Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
  • MDL Number:MFCD00003969
  • Molecular Weight:960.82 g/mol
  • EINECS:200-786-7
Specifications

Specifications

  • VE:
    10 g

Specifications