Pipette tips, Matrix™ SerialMate™
Standard, 0,5 - 30 µl, -, -, 48,26 mm, Matrix™ SerialMate™, 20 racks, 96 each
Catalog # MATR5571
Supplier: THERMO MATRIX TECHNOLOGIES
Jump to:
Product Details & Documents
Pipette tips for use with the Matrix™ SerialMate™ pipetting station. Compatible with 96-well plates.
Specifications
- Verpackt:20 racks, 96 each
- Steril:-
- Volumen:0,5 - 30 µl
- Länge:48,26 mm
- Spitzentyp:Standard
- Zur Nutzung mit:Matrix™ SerialMate™
- Filter:-
- Cat. No.:MATR5571
- Umweltzertifizierung:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Umweltfreundlichkeit:Umweltschonende Herstellung