Cyclohexan ≥99,8%, Laborreagens
Glasflasche, 2,5 L
Anbieter: Honeywell Chemicals
Cyclohexan ≥99,8%, Laborreagens
Artikelnr. C100307-2.5L
Anbieter: Honeywell Chemicals
CAS Nummer: 110-82-7
Springe zu:
Product Details & Documents
- ≥95% for general laboratory use.
Beilstein Number: 1900225
RTECS Number: GU6300000
UNSPSC Code: 12191504
Synonyme: Hexahydrobenzene, Hexamethylene
- CAS Nummer:110-82-7
- Formel:C₆H₁₂
- MDL-Nummer:MFCD00003814
- Molekulargewicht:84.16 g/mol
- Merck-Index:13,02752
- Siedepunkt:8,3 %(V)
- Schmelzpunkt:6,4 °C
- Dichte:0,78 g/cm³ (20 °C)
- Blitzpunkt:–18 °C
- Blitzpunkt:-18 °C
- Lagertemperatur:Raumtemperatur
- EINECS:203-806-2
- UN:1145
- ADR:3,II
Dokumente
Spezifikationen
Spezifikation Testergebnisse
- Appearance (form):complying
- Appearance (color):complying
- Purity (GC):Min. 99.80 Fl.%
- Non-volatile matter:Max. 0.005 %
- IR spectrum:complying
- Vapor Density:2.9 (vs air)
- Vapor Pressure:77 mmHg ( 20 °C)
- Autoignition Temperature:500 °F
- Refractive Index:n20/D 1.426 (lit.)
Spezifikationen
- Packung:Glasflasche
- VE:2,5 L
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



