Ammoniumnitrat, Sigma-Aldrich®
Kunststoffflasche für Feststoffe, 2,5 kg
SIALA3795-2.5KG
: Merck
- :6484-52-2
- :NH₄NO₃
- :MFCD00011425
- :80.04 g/mol
- :13,00538
- :230 °C (1013 hPa)
- :169 °C
- :1,73 g/cm³ (20 °C)
- :Raumtemperatur
- :229-347-8
- :5.1,III
- :M-Clarity™ Qualitätsstufe = MQ200
- :1942
- Packung:Kunststoffflasche für Feststoffe
- VE:2,5 kg
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.