Pharmacy scoops
400 ml, 280 mm, 75 mm
Artikelnr. BOCH8831
Anbieter: BOCHEM
Springe zu:
Product Details & Documents
Stainless steel 18/10.
- Soldered handle
Spezifikationen
- Ø:75 mm
- Kapazität:400 ml
- Länge:280 mm