Express Eingabe
Austria
AuftragsverfolgungKontakt
Pulvertrichter | Avantor
Pulvertrichter
65 mm
Artikelnr. HECH44002030
Pulvertrichter
Artikelnr. HECH44002030
Pulvertrichter
Pulvertrichter
65 mm
Preis
10,70 €
Einloggen für Verfügbarkeit
1 * 5 ST
Product Details & Documents
Powder funnels, made of PP, translucent. For decanting powdery materials. Also suitable for liquids. Cone angle 60°.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Autoclavable
Dokumente
Spezifikationen
  • Trichter A-Ø:
    65 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.