Buffers, concentrated solutions, Fluka™
9,00, ±0.02, sodium tetraborate/hydrochloric acid, 500 ml, Ampoule, 6
Anbieter: Honeywell Chemicals
Buffers, concentrated solutions, Fluka™
Artikelnr. 38748-6X1EA
Anbieter: Honeywell Chemicals
CAS Nummer:
Springe zu:
Product Details & Documents
Documents
Spezifikationen
Spezifikationen
- Packung:Ampoule
- VE:6
- pH Genauigkeit:±0.02
- Endvolumen:500 ml
- pH-Wert:9,00
- Inhalt:sodium tetraborate/hydrochloric acid
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



