Buffers, concentrated solutions, Fluka™
7,00, ±0.02, potassium dihydrogen phosphate/disodium hydrogen phosphate, 500 ml, Ampoule, 1
Anbieter: Honeywell Chemicals
Buffers, concentrated solutions, Fluka™
Artikelnr. 38746-1EA
Anbieter: Honeywell Chemicals
CAS Nummer:
Springe zu:
Product Details & Documents
Dokumente
Spezifikationen
Spezifikationen
- Packung:Ampoule
- VE:1
- pH Genauigkeit:±0.02
- Endvolumen:500 ml
- pH-Wert:7,00
- Inhalt:potassium dihydrogen phosphate/disodium hydrogen phosphate



