Buffers, concentrated solutions, Fluka™
5,00, ±0.02, citric acid/sodium hydroxide solution, 500 ml, Ampoule, 1
Anbieter: Honeywell Chemicals
Buffers, concentrated solutions, Fluka™
Artikelnr. 38744-1EA
Anbieter: Honeywell Chemicals
CAS Nummer:
Springe zu:
Product Details & Documents
Dokumente
Spezifikationen
Spezifikationen
- Packung:Ampoule
- VE:1
- pH Genauigkeit:±0.02
- Endvolumen:500 ml
- pH-Wert:5,00
- Inhalt:citric acid/sodium hydroxide solution
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



