Polystyrene, analytical standards for GPC
~4020 Mn, Mw/Mn ~1,05, 1 g
Artikelnr. 67302-1G
Anbieter: Merck
Springe zu:
Product Details & Documents
Zertifizierung: Exact values can be taken from the accompanying certificate of analysis
Achtung! Bitte beachten: Storage temperature: 2...8 °C. Limited shelf life, expiry date on the label.
Spezifikationen
Spezifikationen
- VE:1 g
- Bezeichnung:~4020 Mn, Mw/Mn ~1,05
- Cat. No.:67302-1G
- M-Clarity Qualitätsstufe:MQ100
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.