Ethyllinoleat
Glass bottle for solids, 100 mg
Catalog # CAYM10008198-100
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: 9Z,12Z-Octadecadienoic acid ethyl ester, Linoleic acid ethyl ester, cis,cis-9,12-Octadecadienoic acid ethyl ester, (9Z,12Z)-Ethyl octadeca-9,12-dienoate
- CAS Number:544-35-4
- Formula:C₂₀H₃₆O₂
- MDL Number:MFCD00009535
- Molecular Weight:308.5 g/mol
- Storage Temperature:Gefrierschrank
- EINECS:208-868-4
Specifications
Specifications
- Packung:Glass bottle for solids
- VE:100 mg



