Micro capillaries, disposable, Kimble®
Capillary pipettes, 10 µl, Orange
Catalog # SCERSP71900-10
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
These disposable micro capillary pipettes are made from soda lime glass, USP Type III.
- Calibrated to contain
- Colour-coded
- Marked with single black capacity ring
- Packaged in easy-open cylinders for convenient dispensing
Certifications: Designed from ASTM Specification E672.
Packaging: Each cylinder contains 250 pipettes and one pipette device.
Specifications
- Farbcode:Orange
- Kapazität:10 µl
- Bezeichnung:Capillary pipettes