Accessories for glove boxes, Precise™
Manual valve gas and vacuum tubing connection kit, with flexible vinyl tubing and fittings necessary to connect user-supplied inert gas supply tank and rotary vane vacuum pump to valves located on the glove box
Artikelnr. LBCP5245100
Anbieter: LABCONCO
Springe zu:
Product Details & Documents
Spezifikationen
- Bezeichnung:Manual valve gas and vacuum tubing connection kit, with flexible vinyl tubing and fittings necessary to connect user-supplied inert gas supply tank and rotary vane vacuum pump to valves located on the glove box
- Seq:02