Human CD62E ELISA Set (without plates)
Human CD62E ELISA Set (without plates), 5×96 tests
Artikelnr. ABCAAB47348-480
Anbieter: Abcam
Springe zu:
Product Details & Documents
Human CD62E ELISA Set (without plates) is a Sandwich (quantitative) ELISA kit for the measurement of Human CD62E ELISA Set (without plates) in Human in Plasma, Cell culture supernatant, Serum samples.
Spezifikationen
Spezifikationen
- Bezeichnung:Human CD62E ELISA Set (without plates)
- Tests pro Kit:5×96 tests
- Empfindlichkeit:0,5 ng/ml
- Reaktivität der primären Antikörper:Human
- Testdauer:Multiple steps
- Time to Results:2 h 15 min
- Cat. No.:ABCAAB47348-480
- Lagertemperatur:4 °C
- Format:Pre-coated
- Target protein:CD62E
- Erkennungsmethode:Colorimetric
- Testtyp:Sandwich
- Probentyp:Plasma, Cell culture supernatant, Serum
- Erfassungsbereich:1 - 32 ng/ml
- Regulatory status:RUO
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



