Express Eingabe
Austria
AuftragsverfolgungKontakt
Mouse IL-2 ELISPOT Set | Avantor

Mouse IL-2 ELISPOT Set

Mouse IL-2 ELISPOT set, 5×96 tests
Artikelnr. ABCAAB46623-480
Anbieter:   Abcam
Mouse IL-2 ELISPOT Set
Artikelnr. ABCAAB46623-480
Anbieter:  Abcam
CAS Nummer:  
Mouse IL-2 ELISPOT Set
Mouse IL-2 ELISPOT Set

Mouse IL-2 ELISPOT set, 5×96 tests

CAS Nummer:  
Preis
1 110,00 €
Einloggen für Verfügbarkeit
1 * 480 Tests
Product Details & Documents

Mouse IL-2 ELISPOT Set is a ELISPOT Set for the sensitive detection of Mouse IL-2 production in a single cell suspension.

Spezifikationen

Spezifikationen

  • Bezeichnung:
    Mouse IL-2 ELISPOT set
  • Tests pro Kit:
    5×96 tests
  • Cat. No.:
    ABCAAB46623-480
  • Lagertemperatur:
    4 °C
  • Species reactivity:
    Mouse
  • Probentyp:
    Suspension cells
  • Product type:
    Uncoated
  • Processing time:
    Multiple steps
  • Antigen-Symbol:
    IL-2
  • Regulatory status:
    RUO

Spezifikationen

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.